missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II (I-A/I-E) Rat, anti-Mouse, FITC, Clone: M5/114.15.2, STEMCELL Technologies
Rat Monoclonal Antibody
POA - £187.65
Specifications
| Antigen | MHC Class II (I-A/I-E) |
|---|---|
| Clone | M5/114.15.2 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17167521
|
Stemcell Technologies
100-0294 |
25 μg | |||||||
17177521
|
Stemcell Technologies
100-0295 |
100 μg |
£187.65
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17187521
|
Stemcell Technologies
100-0296 |
500 μg | |||||||
Description
- The M5/114.15.2 (M5/114) antibody reacts with an extracellular epitope on the mouse major histocompatibility complex class II receptor (MHC-II)
- MHC-II is a heterodimeric transmembrane glycoprotein comprising an α subunit associated non-covalently with a β subunit
- In mice, MHC-II complexes are designated I-A and I-E and comprise Aα/Aβ and Eα/Eβ heterodimers, respectively
- Together with the CD3/T cell receptor (TCR) complex and CD4 molecules, MHC-II mediates a critical function in presenting peptides generated from hydrolysis of exogenous antigens by antigen-presenting cells to CD4+ T (helper) cells, thereby either suppressing or inducing an immune response to the peptides
- The M5/114.15.2 antibody recognizes a polymorphic determinant shared by the I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek (but not I-Af, I-Ak, or I-As) MHC class II alloantigens expressed on B cells, dendritic cells, macrophages, monocytes, and activated T cells of mice carrying the H-2b, d, p, q, r, and u haplotypes (but not haplotypes H-2f or H-2s)
- The M5/114 antibody reportedly inhibits I-A-restricted responses by T cells of the H-2b, d, q, and u (but not H-2f, k, or s) haplotypes
- It does not react with cells from NOD (H-2g7) mice
Specifications
| MHC Class II (I-A/I-E) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FITC | |
| Mouse |
| M5/114.15.2 | |
| Monoclonal | |
| Rat | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title