missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC class II (I-A/I-E) Mouse anti-Mouse, Clone: MK-D6, Novus Biologicals™
Shop All Bio Techne ProductsDescription
MHC class II (I-A/I-E) Monoclonal specifically detects MHC class II (I-A/I-E) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.
Specifications
Specifications
| Antigen | MHC class II (I-A/I-E) |
| Applications | Flow Cytometry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Clone | MK-D6 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Functional |
| Formulation | 10 mM PBS with 0.05% BSA |
| Host Species | Mouse |
| Immunogen | Spleen cells from B10.D2 mice |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?