missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC class II (I-A/I-E) Mouse anti-Mouse, Clone: MK-D6, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | MHC class II (I-A/I-E) |
|---|---|
| Clone | MK-D6 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5-1 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Functional |
| Applications | Flow Cytometry, Immunofluorescence, Functional Assay |
Description
MHC class II (I-A/I-E) Monoclonal specifically detects MHC class II (I-A/I-E) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.Specifications
| MHC class II (I-A/I-E) | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| Spleen cells from B10.D2 mice | |
| Primary | |
| Store at 4°C. |
| MK-D6 | |
| Flow Cytometry 0.5-1 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human (Negative), Mouse | |
| 100504404 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title