missing translation for 'onlineSavingsMsg'
Learn More

MHC class II (I-A/I-E) Antibody (M5/114.15.2), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18419734 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419734 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419734 Supplier Novus Biologicals Supplier No. NBP143312AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

MHC class II (I-A/I-E) Monoclonal antibody specifically detects MHC class II (I-A/I-E) in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC class II (I-A/I-E)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M5/114.15.2
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias E-alpha-f;H-2 class II histocompatibility antigen, E-K alpha chain;H-2 class II histocompatibility antigen, E-U alpha chain;H-2E;H2-Ea;H-2Ea;H2-Ea-ps;I;Ia;Ia3;Ia-3;I-E;I-Ealpha;MHC-H2-Ea
Host Species Rat
Immunogen The immunogen for this antibody was MHC Class II.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 100504404
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.