missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II antigen I A beta 1 Antibody (25-9-3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28162
This item is not returnable.
View return policy
Description
MHC Class II antigen I A beta 1 Monoclonal specifically detects MHC Class II antigen I A beta 1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| MHC Class II antigen I A beta 1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| HLA-DRB1 | |
| C3H.SW mouse splenocytes | |
| 0.5 mg | |
| Asthma, Immunology, Major Histocompatibility Complexes | |
| 3123 | |
| Store at 4C. Do not freeze. | |
| IgM |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 25-9-3 | |
| Unconjugated | |
| Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly | |
| Mouse | |
| Multi-step | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction