missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Antibody (NIMR-4), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-28159
This item is not returnable.
View return policy
Description
MHC Class II Monoclonal specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
| MHC Class II | |
| Monoclonal | |
| 0.5mg/mL | |
| Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry | |
| HLA-DMA | |
| The immunogen is a peptide made to the amino acid region MHC Class II | |
| 0.25 mg | |
| Immunology, Major Histocompatibility Complexes | |
| 3108 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| NIMR-4 | |
| FITC | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| MHC Class II molecules | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction