missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MHC Class II Monoclonal specifically detects MHC Class II in Human, Bovine, Canine, Equine, Feline, Goat, Hamster, Llama, Alpaca, Bison, Ferret, Mink, Water Buffalo, Rabbit, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
| Antigen | MHC Class II |
| Applications | Flow Cytometry, Immunohistochemistry, Functional Assay |
| Classification | Monoclonal |
| Clone | H42A |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:10 - 1:1000 |
| Formulation | Ascites with 0.09% Sodium Azide |
| Gene Alias | class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM |
| Gene Symbols | HLA-DMA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?