Learn More
MHC Class II Antibody (CVS20), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34848H
Product Code. 18447011
  • £570.00 / 0.10mL
Description
MHC Class II Monoclonal antibody specifically detects MHC Class II in Human, Bovine, Canine, Equine samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MHC Class II
Monoclonal
HRP
PBS
Mouse
Protein G purified
RUO
Primary
Human, Bovine, Canine, Equine
Purified
Immunohistochemistry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
CVS20
Immunohistochemistry
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
3132 cells
0.1 mL
Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
3108
Store at 4C in the dark.
IgG1