missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I H-2Kk Antibody (16-3-22S) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28202
This item is not returnable.
View return policy
Description
MHC Class I H-2Kk Monoclonal specifically detects MHC Class I H-2Kk in Mouse samples. It is validated for Flow Cytometry.
Specifications
| H-2Kk | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| H2-K1 | |
| C3H mouse splenocytes | |
| 0.25 mg | |
| Primary | |
| An epitope on mouse H-2Kk MHC class I alloantigen | |
| Store at 4C. Do not freeze. | |
| IgG2a κ |
| Flow Cytometry, Immunoprecipitation | |
| 16-3-22S | |
| Unconjugated | |
| adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 14972 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction