Learn More
MHC Class I (H-2Kb) Monoclonal Antibody (AF6-88.5.5.3), eFluor™ 450, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£70.00 - £232.00
Specifications
| Antigen | MHC Class I (H-2Kb) |
|---|---|
| Clone | AF6-88.5.5.3 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Description: This AF6-88.5.5.3 monoclonal antibody reacts with the H-2Kb MHC class I alloantigen. H-2Kb is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. d, f, j, k, p, q, r, s, u, and v) has not been observed. Applications Reported: This AF6-88.5.5.3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AF6-88.5.5.3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| MHC Class I (H-2Kb) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 14972 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| AF6-88.5.5.3 | |
| Flow Cytometry | |
| eFluor 450 | |
| Mouse | |
| Mouse | |
| H2-K1 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| H2-K1 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.