missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MHC Class I (H-2Db) Monoclonal Antibody (28-14-8), NovaFluor™ Yellow 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£154.00 - £297.00
Specifications
| Antigen | MHC Class I (H-2Db) |
|---|---|
| Clone | 28-14-8 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30054834
|
Invitrogen™
M038T03Y08-A |
100 μg |
£297.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055971
|
Invitrogen™
M038T02Y08-A |
25 μg |
£154.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MHC Class I (H-2Db) Monoclonal antibody specifically detects MHC Class I (H-2Db) in Mouse samples. It is validated for Flow CytometrySpecifications
| MHC Class I (H-2Db) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Mouse | |
| Mouse | |
| P01895, P01896, P01897, P01899, P01900, P14426, P14427 | |
| 14964 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 28-14-8 | |
| Flow Cytometry | |
| NovaFluor Yellow 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2DB; H2-DC1-beta transplantation antigen protein; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein | |
| IgG2a κ | |
| Affinity chromatography | |
| H2-D1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title