missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (R1-21.2) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£225.00 - £477.00
Specifications
| Antigen | MHC Class I |
|---|---|
| Clone | R1-21.2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmunoassay |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667116
|
Novus Biologicals
NBP3-09017-0.2mg |
0.2 mg |
£477.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18087725
|
Novus Biologicals
NBP3-09017-0.025MG |
0.025 mg |
£225.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MHC Class I Monoclonal antibody specifically detects MHC Class I in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune AssaysSpecifications
| MHC Class I | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Stem Cell Markers | |
| PBS | |
| 3133 | |
| IgG κ | |
| Protein A purified |
| R1-21.2 | |
| Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmunoassay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| This antibody was raised by immunising Lewis 1F rats with mouse CH-1 lymphoma cells. Spleen cells from immunised rats were fused with X-63Ag8-653 myeloma cells to generate hybridomas. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title