missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (MEM-E/08), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-305APCCY7
This item is not returnable.
View return policy
Description
MHC Class I Monoclonal specifically detects MHC Class I in Human samples. It is validated for Flow Cytometry.
Specifications
| MHC Class I | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| HLA-E | |
| Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide. | |
| 0.1 mL | |
| Primary | |
| This antibody (clone MEM-E/08) recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The results revealed that antibody is remarkably specific for HLA-E, only with weak cross-reactivity with following classical MHC Class I molecules (MHC Class Ia): HLA-A24, -B7, -B27, -B51, -B54, -C7. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| MEM-E/08 | |
| Flow Cytometry | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Major Histocompatibility Complexes, Stem Cell Markers | |
| 3133.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction