missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (MEM-E/06), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-307JF549
This item is not returnable.
View return policy
Description
MHC Class I Monoclonal specifically detects MHC Class I in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MHCClassI | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HLA-E | |
| Bacterially expressed recombinant full length protein refolded with beta2-microglobulin and peptide (Human). | |
| 0.1 mL | |
| Primary | |
| This antibody (clone MEM-E/06) recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. Some resultss showed that this antibody cross-reacts with some classical MHC Class I molecules (HLA-A3, -A11, -B7). However, others have confirmed that this antibody exhibits much broader cross-reactivity classical MHC Class I antigens, namely with HLA-A24, -A32, -B8, -B15, -B27, -B35, -B44, -B54, -C3, -C4, -C5, -C7. | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MEM-E/06 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Major Histocompatibility Complexes, Stem Cell Markers | |
| 3133 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction