missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MGST3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140297
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: INVSKARKKY KVEYPIMYST DPENGHIFNC Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 90%.
The MAPEG family consists of six human proteins, several of which are involved the production of leukotrienes and prostaglandin E, important mediators of inflammation. This gene encodes an enzyme which catalyzes the conjugation of leukotriene A4 and reduced glutathione to produce leukotriene C4. This enzyme also demonstrates glutathione-dependent peroxidase activity towards lipid hydroperoxides.
Specifications
| MGST3 | |
| Polyclonal | |
| Unconjugated | |
| MGST3 | |
| 2010012L10Rik; 2010306B17Rik; 2700004G04Rik; AA516734; Glutathione peroxidase MGST3; GST-III; LTC4 synthase MGST3; MGST3; microsomal glutathione S-transferase; microsomal glutathione S-transferase 3; Microsomal glutathione S-transferase III; microsomal GST-3; Microsomal GST-III | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4259 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14880 | |
| MGST3 | |
| Recombinant protein corresponding to Human MGST3. Recombinant protein control fragment (Product #RP-109660). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction