missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGST2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98515H
This item is not returnable.
View return policy
Description
MGST2 Polyclonal antibody specifically detects MGST2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MGST2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 4258 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| HRP | |
| EC 2.5.1.18, FLJ27438, GST2MGC14097, MGST-II, microsomal glutathione S-transferase 2, Microsomal GST-2, Microsomal GST-II, truncated microsomal glutathione S-transferase 2 | |
| Produced in rabbits immunized with E. coli-derived Human MGST2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction