missing translation for 'onlineSavingsMsg'
Learn More

MGRN1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18359964 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359964 100 μg 100µL
18340624 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18359964 Supplier Novus Biologicals Supplier No. NBP317891100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MGRN1 Polyclonal antibody specifically detects MGRN1 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGRN1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias E3 ubiquitin-protein ligase MGRN1, EC 6.3.2, KIAA0544probable E3 ubiquitin-protein ligase MGRN1, Mahogunin RING finger protein 1, mahogunin, ring finger 1, RING finger protein 156, RNF156EC 6.3.2.-
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: LPFRALLQIRAVRKKPGALSPVSFSPVLAQSLEHDEHSNSDSVPPGYEPISLLEALNGLRAVSPAIPSAPLYEEITYSGISDGLSQASCPLAAIDHILDSSRQKGRPQSKAPDSTLRSPSSPIHEEDE
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 23295
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.