missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ MGM™-4 Melanocyte Growth Medium-4 BulletKit™

Product Code. 11625480
Click to view available options
:
Medium: 500 mL, CaCl2: 1.0 mL, PMA: 0.50 mL, hFGF-B: 1.0 mL, Insulin: 1.0 mL, BPE: 2.0 mL, Hydrocortisone: 0.50 mL, FBS: 2.50 mL, GA-1000: 0.50 mL
This item is not returnable. View return policy

Product Code. 11625480

Brand: Lonza™ CC3249

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Complete kit, growth medium and supplements, to support growth of melanocytes.

  • Culture kit containing growth medium and supplements.
  • Developed to support optimal growth of melanocytes.
  • Provided in a BulletKit™ format for increased flexibility to add or omit components as research needs dictate.
  • Includes the following:
    • 1 x MBM™-4 Basal Medium (CC-3250), 500 mL
    • MGM™-4 SingleQuots™ Supplement Pack (CC-4435) containing the following:
      • 1 x Natural Cap Vial with CaCl2, 1.00 mL
      • 1 x Amber Vial with PMA, 0.50 mL
      • 1 x Gray Cap Vial with hFGF-B, 1.00 mL
      • 1 x Lilac Cap Vial with Insulin, 1.00 mL
      • 1 x Orange Cap Vial with BPE, 2.00 mL
      • 1 x Natural Cap Vial with Hydrocortisone, 0.50 mL
      • 1 x Bottle FBS, 2.50 mL
      • 1 x Red Cap Vial with GA-1000, 0.50 mL
  • ET-3 Growth Supplement an additional growth factor required for adult melanocyte growth, is also sold separately to supplement the MGM™-4 BulletKit™ for a complete formulation specifically designed for growing adult melanocytes.
  • ET-3 offers enhanced growth and plating advantages in addition to limiting other cell type contamination in culture.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Melanocytes
Product Type Growth Medium and Supplements
Quantity Medium: 500 mL, CaCl2: 1.0 mL, PMA: 0.50 mL, hFGF-B: 1.0 mL, Insulin: 1.0 mL, BPE: 2.0 mL, Hydrocortisone: 0.50 mL, FBS: 2.50 mL, GA-1000: 0.50 mL
pH 7.25 to 7.65
Color Red
Content And Storage Growth Medium, CaCl2, PMA, hFGF-B, Insulin, BPE, Hydrocortisone, FBS, GA-1000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.