missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR8 Antibody (4C1C1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£184.00 - £453.00
Specifications
| Antigen | mGluR8 |
|---|---|
| Clone | 4C1C1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18233054
|
Novus Biologicals
NBP2-61857 |
0.1 mL |
£453.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654727
|
Novus Biologicals
NBP2-61857-0.025ml |
0.025 mL |
£184.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mGluR8 Monoclonal specifically detects mGluR8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| mGluR8 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| FLJ41058, glutamate metabotropic receptor 8, glutamate receptor, metabotropic 8, GPRC1HMGLUR8GLUR8, metabotropic glutamate receptor 8, MGC126724, mGlu8, mGluR8 | |
| GRM8 | |
| IgG1 | |
| Protein G purified |
| 4C1C1 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2918 | |
| Purified recombinant fragment of human GRM8 (AA: extra 440-583) expressed in E. Coli. | |
| Primary | |
| 101.7 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title