missing translation for 'onlineSavingsMsg'
Learn More

mGluR8 Antibody (4A7), Novus Biologicals™

Product Code. 18369998 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369998 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369998 Supplier Novus Biologicals Supplier No. H00002918M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

mGluR8 Monoclonal antibody specifically detects mGluR8 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR8
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4A7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000836
Gene Alias FLJ41058, glutamate metabotropic receptor 8, glutamate receptor, metabotropic 8, GPRC1HMGLUR8GLUR8, metabotropic glutamate receptor 8, MGC126724, mGlu8, mGluR8
Host Species Mouse
Immunogen GRM8 (NP_000836, 486 a.a. ~ 575 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KVIGHWTNQLHLKVEDMQWAHREHTHPASVCSLPCKPGERKKTVKGVPCCWHCERCEGYNYQVDELSCELCPLDQRPNMNRTGCQLIPII
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2918
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.