missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR7 Antibody (7G8D3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £435.00
Specifications
| Antigen | mGluR7 |
|---|---|
| Clone | 7G8D3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18265511
|
Novus Biologicals
NBP2-61856 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671397
|
Novus Biologicals
NBP2-61856-0.025ml |
0.025 mL |
£173.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mGluR7 Monoclonal specifically detects mGluR7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| mGluR7 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| FLJ40498, glutamate receptor, metabotropic 7, metabotropic glutamate receptor 7, mGlu7, mGluR7, MGLUR7GPRC1GGLUR7 | |
| GRM7 | |
| IgG1 | |
| Protein G purified |
| 7G8D3 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision | |
| 2917 | |
| Purified recombinant fragment of human GRM7 (AA: 454-590) expressed in E. Coli. | |
| Primary | |
| 102.3 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title