missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528770
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: Ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR17, NMDA13, KAI2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.
Specifications
| mGluR6 | |
| Polyclonal | |
| Unconjugated | |
| GRM6 | |
| BC021919; CSNB1B; G protein-coupled receptor GRM6; G protein-coupled receptor, family C, group 1, member F; glutamate metabotropic receptor 6; glutamate receptor; glutamate receptor, metabotropic 6; Gm3; GPRC1F; Grm6; Metabotropic glutamate receptor 6; mGlu6; MGLUR6; nerg1; nob2; nob3; nob4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2916 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O15303 | |
| GRM6 | |
| Synthetic peptide corresponding to a region within amino acids 813 and 877 of Human Metabotropic Glutamate Receptor 6 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction