missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mGluR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550999
This item is not returnable.
View return policy
Description
mGluR4 Polyclonal Antibody detects endogenous levels of mGluR4 protein.
The Metabotropic Glutamate Receptor mGluR4/GRM4 binds L-glutamate, the major excitatory neurotransmitter in the central nervous system, which activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties.
Specifications
| mGluR4 | |
| Polyclonal | |
| Unconjugated | |
| GRM4 | |
| G protein-coupled receptor GRM4; G protein-coupled receptor, family C, group 1, member D; glutamate metabotropic receptor 4; glutamate receptor; glutamate receptor, metabotropic 4; GPRC1D; GRM4; Metabotropic glutamate receptor 4; mGlu4; MGLUR4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24417, 268934, 2914 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31423, Q14833, Q68EF4 | |
| GRM4 | |
| Synthesized peptide derived from the C-terminal region of human mGluR-4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction