missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mGluR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533820
This item is not returnable.
View return policy
Description
mGluR3 Polyclonal Antibody for ICC/IF, IHC (P)
Glutamate is the main excitatory neurotransmitter in the brain. For many years it had been considered to act only on the ligand-gated receptor channels-termed NMDA, AMPA and kainite receptors that are involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to regulate enzymes producing second messengers via specific receptors coupled to G-proteins. These receptors are called metabotropic glutamate receptors. In expression systems, Group-I receptors stimulate phospholipase C as revealed by an increase in phosphoinositide turnover and calcium release from internal stores. Group-II and -III receptors are coupled to the inhibition of adenylyl cyclase. The Group-I receptors include mGluR1a and mGluR5. The Group-II receptors in include mGluR2 and mGluR3.
Specifications
| mGluR3 | |
| Polyclonal | |
| Unconjugated | |
| GRM3 | |
| 0710001G23Rik; G protein-coupled receptor, family C, group 1, member C; GLUR3; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor, metabotropic 3; Gprc1c; GRM3; M mGlu3; Metabotropic glutamate receptor 3; mGlu3; mGluR3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100049918, 100521916, 100760036, 108069, 24416, 2913, 541056 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P31422, Q14832, Q9QYS2 | |
| GRM3 | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GRM3 / MGLUR3. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine, Hamster, Pig, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction