missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
mGluR3 Monoclonal antibody specifically detects mGluR3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | mGluR3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SR1543 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | GLUR3, glutamate receptor, metabotropic 3, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human mGluR3 (Uniprot #: Q14832) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?