missing translation for 'onlineSavingsMsg'
Få mere at vide

mGluR2 Rabbit anti-Mouse, Rat, Clone: 0U6B5, Novus Biologicals™

Artikelnummer. 18359314 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
100 μg
20 μg
Pakningsstørrelse:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18359314 100 μg 100µL
18308576 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18359314 Leverandør Novus Biologicals Leverandørnr. NBP316243100UL

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Monoclonal Antibody

mGluR2 Monoclonal antibody specifically detects mGluR2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen mGluR2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 0U6B5
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1Bmetabotropic glutamate receptor 2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 773-872 of human mGluR2 (Q14416).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2912
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.