Learn More
Invitrogen™ mGluR1 Recombinant Rabbit Monoclonal Antibody (JM11-61)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532635
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Glutamate is the main excitatory neurotransmitter in the brain. For many years it had been considered to act only on the ligand-gated receptor channels-termed NMDA, AMPA and kainite receptors that are involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to regulate enzymes producing second messengers via specific receptors coupled to G-proteins. These receptors are called metabotropic glutamate receptors. In expression systems, Group-I receptors stimulate phospholipase C as revealed by an increase in phosphoinositide turnover and calcium release from internal stores. Group-II and -III receptors are coupled to the inhibition of adenylyl cyclase. The Group-I receptors include mGluR1a and mGluR5. The Group-II receptors in include mGluR2 and mGluR3.
Specifications
| mGluR1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P23385, P97772, Q13255 | |
| Grm1 | |
| Recombinant protein within Human mGluR1 aa 280-420. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| JM11-61 | |
| Unconjugated | |
| Grm1 | |
| 4930455H15Rik; ENSMUSG00000075319; G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member A; glutamate metabotropic receptor 1; glutamate receptor; glutamate receptor, metabotropic 1; Gm10828; GPRC1A; GRM1; GRM1A; Metabotropic glutamate receptor 1; MGLU1; MGLUR1; mGluR1a; nmf373; PPP1R85; protein phosphatase 1, regulatory subunit 85; rcw; SCAR13; wobl | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14816, 24414, 2911 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.