missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR1 Antibody (5E12D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61827-0.025ml
This item is not returnable.
View return policy
Description
mGluR1 Monoclonal specifically detects mGluR1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| mGluR1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 | |
| Mouse | |
| 132.4 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 5E12D8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GRM1 | |
| Purified recombinant fragment of human GRM1 (AA: extra 387-486) expressed in E. Coli. | |
| Protein G purified | |
| GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 2911 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction