missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGEA5, Mouse anti-Human, Clone: 1C7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010724-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: SFHEEQEVLPETFLANFPSLIKMDIHKKVTDPSVAKSMMACLLSSLKANGSRGAFCEVRPDDKRILEFYSKLGCFEIAKMEGFPKDVVILGRSLSpecifications
| MGEA5 | |
| Monoclonal | |
| Unconjugated | |
| meningioma expressed antigen 5 (hyaluronidase) | |
| FLJ11229/FLJ23355/KIAA0679/MEA5/NCOAT/OGA | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 1C7 | |
| In 1x PBS, pH 7.4 | |
| NM_012215 | |
| MGEA5 | |
| MGEA5 (NP_036347, 823 a.a. ∼ 916 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10724 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction