missing translation for 'onlineSavingsMsg'
Learn More

MgcRacGAP/RACGAP1 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18455515 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18455515 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18455515 Supplier Novus Biologicals Supplier No. NBP147267UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MgcRacGAP/RACGAP1 Polyclonal antibody specifically detects MgcRacGAP/RACGAP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MgcRacGAP/RACGAP1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias GTPase activating protein, HsCYK-4, ID-GAP, KIAA1478, Male germ cell RacGap, MGCRACGAP, MgcRacGAPrac GTPase-activating protein 1, Rac GTPase activating protein 1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 150 and 200 of human Rac GTPase-Activating Protein 1 using the numbering given in entry NP_001119575.1 (GeneID 29127).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 29127
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.