missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ MgCl2 10 mM Solution

Product Code. 11670441
Change view
Click to view available options
:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11670441 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11670441 Supplier Lonza™ Supplier No. S50641

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used as the sample diluent when attempting to overcome inhibitory chelation effect.

  • Can be used as the sample diluent when attempting to overcome inhibitory chelation effect.
  • Examples of samples which chelate divalent cations include heparin and EDTA.
  • May be used to prepare samples for any LAL assay.
  • Contains <0.005 EU/mL endotoxin.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material MgCl2
Description Contains <0.005 EU/mL endotoxin
Concentration 10 mM
For Use With (Application) Standard PCR
Quantity 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.