missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGCD-265 analog, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Description
MGCD-265 analog is a potent and oral active inhibitor of c-Met and VEGFR2 tyrosine kinases, with IC50s of 29 nM and 10 nM, respectively. MGCD-265 analog has significant antitumor activity.
Specifications
Specifications
| Chemical Name or Material | MGCD-265 analog |
| CAS | 875337-44-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H20FN5O2S2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (193.20 mM) |
| SMILES | O=C(CC1=CC=CC=C1)NC(NC2=CC=C(C(F)=C2)OC3=C4C(C=C(S4)C5=CN(C=N5)C)=NC=C3)=S |
| Molecular Weight (g/mol) | 517.6 |
| Formula Weight | 517.6 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction