missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGC4172 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35434-100ul
This item is not returnable.
View return policy
Description
MGC4172 Polyclonal antibody specifically detects MGC4172 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| MGC4172 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ARPG836, dehydrogenase/reductase (SDR family) member 11, dehydrogenase/reductase SDR family member 11, FLJ39232, MGC4172, SDR24C1, short chain dehydrogenase/reductase family 24C, member 1, short-chain dehydrogenase/reductase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human MGC4172 (NP_077284.2).,, Sequence:, MARPGMERWRDRLALVTGASGGIGAAVARALVQQGLKVVGCARTVGNIEELAAECKSAGYPGTLIPYRCDLSNEEDILSMFSAIRSQHSGVDICINNAGLARPDTLLSGSTSGWKDMFNVNVLALSICTREAYQSMKERNVDDGHIININSMSGHRVLPLSVTHFYSATKYAVTALTEGLRQELREAQTHIRATCISPGVVETQFAFKLHDKDPEKAAATYEQMKCLKPEDVAEAVIYVLSTPAHIQIGDIQMRPTEQVT | |
| 100 μL | |
| Stem Cell Markers | |
| 79154 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction