missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGC20410 Antibody (PCRP-BATF2-2B9), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £385.00
Specifications
| Antigen | MGC20410 |
|---|---|
| Clone | PCRP-BATF2-2B9 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 μg/million cells, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644864
|
Novus Biologicals
NBP3-20601-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657164
|
Novus Biologicals
NBP3-20601-100ug |
100 μg |
£385.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MGC20410 Monoclonal antibody specifically detects MGC20410 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| MGC20410 | |
| 0.2 mg/ml | |
| Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| basic leucine zipper transcription factor, ATF-like 2, basic leucine zipper transcriptional factor ATF-like 2, B-ATF-2, MGC20410, SARI, suppressor of AP-1 regulated by IFN | |
| Recombinant fragment (around aa5-140) of human MGC20410 (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
| PCRP-BATF2-2B9 | |
| Flow Cytometry 1-2 μg/million cells, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA | |
| 116071 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title