missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGC20410 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | MGC20410 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MGC20410 Polyclonal specifically detects MGC20410 in Human samples. It is validated for Western Blot.Specifications
| MGC20410 | |
| Polyclonal | |
| Rabbit | |
| NP_612465 | |
| 116071 | |
| Synthetic peptide directed towards the N terminal of human BATF2. Peptide sequence MHLCGGNGLLTQTDPKEQQRQLKKQKNRAAAQRSRQKHTDKADALHQQHE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| basic leucine zipper transcription factor, ATF-like 2, basic leucine zipper transcriptional factor ATF-like 2, B-ATF-2, MGC20410, SARI, suppressor of AP-1 regulated by IFN | |
| BATF2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title