missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MGAT5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569575
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.
Glycosyltransferase that acts on alpha-linked mannose of N-glycans and O-mannosyl glycans. Catalyzes the transfer of N-acetylglucosamine (GlcNAc) to the beta 1-6 linkage of the mannose residue of GlcNAcbeta1,2-Manalpha on both the alpha1,3- and alpha1,6-linked mannose arms in the core structure of N-glycan. Also acts on the GlcNAcbeta1,2-Manalpha1-Ser/Thr moiety, forming a 2,6-branched structure in brain O-mannosyl glycan. Plays an active role in modulating integrin and laminin-dependent adhesion and migration of neuronal cells via its activity in the O-mannosyl glycan pathway.
Specifications
| MGAT5B | |
| Polyclonal | |
| Unconjugated | |
| MGAT5B | |
| Alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase B; alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase B; beta(1,6)-N-acetylglucosaminyltransferase V; beta1,6-N-acetylglucosaminyltransferase IX (GnT-IX); C330018B01; GlcNAc-T Vb; GnT-IX; GNT-Vb; hGnTVb; KIAA2008; mannoside acetylglucosaminyltransferase 5, isoenzyme B; mannoside acetylglucosaminyltransferase 5B; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isoenzyme B; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B; Mgat5b; mGnTVB; N-acetylglucosaminyltranferase VB; N-acetylglucosaminyltransferase IX; N-acetylglucosaminyl-transferase Vb; UDP-N-acetylglucosamine: alpha1,6-D-mannoside beta1,6-N-acetylglucosaminyltransferase b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 303693 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| MGAT5B | |
| synthetic peptide directed towards the middle region of human MGAT5B. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction