missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MGAT5 Recombinant Rabbit Monoclonal Antibody (7W10I10)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557862
This item is not returnable.
View return policy
Description
Immunogen sequence: CHGQVMWPPL SALQVKLAEP GQSCKQVCQE SQLICEPSFF QHLNKDKDML KYKVTCQSSE LAKDILVPSF DPKNKHCVFQ GDLLLFSCAG AHPRHQRVCP CRDFIKGQVA LCKDCL.
Catalyzes the addition of N-acetylglucosamine in beta 1- 6 linkage to the alpha-linked mannose of biantennary N-linked oligosaccharides. It is one of the most important enzymes involved in the regulation of the biosynthesis of glycoprotein oligosaccharides.
Specifications
| MGAT5 | |
| Recombinant Monoclonal | |
| 0.55 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q08834, Q09328, Q8R4G6 | |
| MGAT5 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7W10I10 | |
| Unconjugated | |
| MGAT5 | |
| 4930471A21Rik; 5330407H02Rik; AI480971; alpha-1,6-mannosyl-glycoprotein 6-beta-N-acetylglucosaminyltransferase; alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase V; beta1,6N-acetylglucosaminyltransferase V; EC 2.4.1.155; GGNT5; GlcNAc-T V; GlcNAc-TV; GNT-V; GNT-VA; GNT-VGNT-VA; mannoside acetylglucosaminyltransferase 5; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase; Mgat5; N-acetylglucosaminyltransferase V; N-acetylglucosaminyl-transferase V; Secreted alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Secreted beta-1,6-N-acetylglucosaminyltransferase V; Secreted GNT-V | |
| Rabbit | |
| Protein A | |
| RUO | |
| 107895, 4249, 65271 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction