Learn More
Invitrogen™ MGAT4C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589756
Description
Immunogen sequence: KNVIRFKPSL FQHMGYYSSY KGTENKLKDD DFEEESFDIP DNPPASLYTN MNVFENYEAS KAYSSVDEYF WGKPPSTGDV FVIVFENPII IKKIKVNTGT EDRQNDILHH GALDVGENVM PSKQRRQCST YLRLGEFKNG NFEMSGVNQK IPFDIHCMRI YVTKTQKEWL IIRSISIWTS; Positive Samples: U-87MG; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
Glycosyltransferase that participates in the transfer of N-acetylglucosamine (GlcNAc) to the core mannose residues of N- linked glycans. Catalyzes the formation of the GlcNAcbeta1-4 branch on the GlcNAcbeta1-2Manalpha1-3 arm of the core structure of N-linked glycans. Essential for the production of tri- and tetra-antennary N-linked sugar chains. Does not catalyze the transfer of GlcNAc to the Manalpha1-6 arm to form GlcNAcBeta1-4Manalpha1-6 linkage ('GnT-VI' activity).
Specifications
| MGAT4C | |
| Polyclonal | |
| Unconjugated | |
| MGAT4C | |
| 9130411I17Rik; Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C; glcNAc-T IVc; GnT-IVc; GntIVh; hGnT-IV-H; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C (putative); MGAT4 family member C; MGAT4 family, member C; MGAT4C; N-acetylglucosaminyltransferase IV homolog; N-acetylglucosaminyltransferase IVc; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVc; RGD1307871; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVc; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV-homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25834 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UBM8 | |
| MGAT4C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 299-478 of human MGAT4C (NP_0373762). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.