Learn More
MGAT4B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30718-1-AP-20UL
Description
The MGAT4B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MGAT4B. This antibody recognizes human, mouse antigen. The MGAT4B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| MGAT4B | |
| 220 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| GlcNAc T IVb, GNT IV, GNT IVB, MGAT4B | |
| 20 μL | |
| 11282 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |