missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MGAT4B Polyclonal antibody specifically detects MGAT4B in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | MGAT4B |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_463459.1 |
| Gene Alias | alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase B, alpha-1,3-mannosyl-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, aminyltransferase, EC 2.4.1.145, GlcNAc-T IVb, GNT-IV, GNT-IVB, isoenzyme B, isozyme B, mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, N-acetylglucosaminyltransferase IVb, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVb, UDP-N-acetylglucosamine: alpha-1,3-D-mannosidebeta-1,4-N-acetylglucosaminyltransferase IV, UDP-N-acetylglucosamine: alpha-1,3-D-mannosidebeta-1,4-N-acetylglucosaminyltransferase IVb |
| Host Species | Mouse |
| Immunogen | MGAT4B (NP_463459.1, 1 a.a. - 563 a.a.) full-length human protein. MSRVAGTRTDVNELLQRWTPRCVRWHTGGARRVALDRPLVTACLPPAGDVVDVYQREFLALRDRLHAAEQESLKRSKELNLVLDEIKRAVSERQALRDGDGNRTWGRLTEDPRLKPWNGSHRHVLHLPTVFHHLPHLLAKESSLQPAVRVGQGRTGVSVVMGIPSVRREVHSYLTDTLHSLISELSPQEKEDSVIVVLIAETDSQYTSAVTENIKALFPTEIHSGLLEVISPSPHFYPDFSRLRESFGDPKERVRWRTKQNLDYCFLMMYAQSKGIYYVQLEDDIVAKPNYLSTMKNFALQQPSEDWMILEFSQLGFIGKMFKSLDLSLIVEFILMFYRDKPIDWLLDHILWVKVCNPEKDAKHCDRQKANLRIRFKPSLFQHVGTHSSLAGKIQKLKDKDFGKQALRKEHVNPPAEVSTSLKTYQHFTLEKAYLREDFFWAFTPAAGDFIRFRFFQPLRLERFFFRSGNIEHPEDKLFNTSVEVLPFDNPQSDKEALQEGRTATLRYPRSPDGYLQIGSFYKGVAEGEVDPAFGPLEALRLSIQTDSPVWVILSEIFLKKAD |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?