missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MGAT3 Polyclonal Antibody

Description
Recommended positive controls: HeLa, HepG2. Predicted reactivity: Mouse (98%), Rat (98%), Chicken (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. MGAT3 (N-acetylglucosaminyltransferase III) transfers a GlcNAc residue to the beta-linked mannose of the trimannosyl core of N-linked oligosaccharides and produces a bisecting GlcNAc. Expression of this gene may be controlled by a multiple-promoter system.
Specifications
Specifications
| Antigen | MGAT3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.96 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | MGAT3 |
| Gene Accession No. | Q09327, Q10470 |
| Gene Alias | 1110038J12Rik; AI848272; Beta-1,4-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase; GGNT3; GlcNAc-T III; GNT3; GNT-III; mannoside acetyl glucosaminyltransferase 3; mannoside acetylglucosaminyltransferase 3; mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase; MGAT3; N-acetylglucosaminyltransferase III; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase III; RP5-1104E15.2 |
| Gene Symbols | MGAT3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?