missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGAT3, Mouse anti-Human, Clone: 2G4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004248-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LVSAQNGDFPRWGDYEDKRDLNYIRGLIRTGGWFDGTQQEYPPADPSEHMYAPKYLLKNYDRFHYLLDNPYQEPRSTAAGGWRHRGPEGRPPARGKLDEAEVSpecifications
| MGAT3 | |
| Monoclonal | |
| Unconjugated | |
| mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase | |
| FLJ43371/GNT-III/GNT3/MGC141943/MGC142278 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 2G4 | |
| In 1x PBS, pH 7.4 | |
| NM_002409 | |
| MGAT3 | |
| MGAT3 (NP_002400, 430 a.a. ∼ 531 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4248 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction