missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MGAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566042
This item is not returnable.
View return policy
Description
Immunogen sequence: QVFFPFSIQLY PNEFPGSDPR DCPRDLPKNA ALKLGCINAE YPDSFGHYRE AKFSQTKHHW WWKLHFVWER Highest antigen sequence identity to the following orthologs - mouse 99%, rat 97%.
The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.
Specifications
| MGAT2 | |
| Polyclonal | |
| Unconjugated | |
| MGAT2 | |
| AA407964; alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase-like protein; beta-1,2-N-acetylglucosaminyltransferase II; beta-12-N-acetylglucosaminyltransferase II; CDG2A; CDGS2; CH73-301C19.1; GlcNAc-T II; GLCNACTII; GNT2; GNT-II; Mannoside acetylglucosaminyltransferase 2; mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase; mgat2; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II; zgc:162268 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4247 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q10469 | |
| MGAT2 | |
| Recombinant Human MGAT2. Recombinant protein control fragment (Product #RP-106732). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction