missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGAT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | MGAT2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263864
|
Novus Biologicals
NBP2-58625 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664137
|
Novus Biologicals
NBP2-58625-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MGAT2 Polyclonal specifically detects MGAT2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| MGAT2 | |
| Polyclonal | |
| Rabbit | |
| metabolism, Proteases & Other Enzymes | |
| alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase, Beta-1,2-N-acetylglucosaminyltransferase II, CDG2A, EC 2.4.1.143, GlcNAc-T II, GLCNACTII, GNT2, GNT-IICDGS2, Mannoside acetylglucosaminyltransferase 2, mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II, UDP-N-acetylglucosamine:alpha-6-D-mannosidebeta-1,2-N-acetylglucosaminyltransferase II | |
| MGAT2 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 4247 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QVFFPFSIQLYPNEFPGSDPRDCPRDLPKNAALKLGCINAEYPDSFGHYREAKFSQTKHHWWWKLHFVWER | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title