missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGARP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543494
This item is not returnable.
View return policy
Description
Peptide sequence: EAVTIDNDKD TTKNETSDEY AELEEENSPA ESESSAGDDL QEEASVGSEA Sequence homology: Human: 100%
MGARP Polyclonal antibody specifically detects MGARP in Human samples. It is validated for Western Blot.
Specifications
| MGARP | |
| Polyclonal | |
| Unconjugated | |
| MGARP | |
| C4orf49; CESP1; CESP-1; Corneal endothelium-specific protein 1; GS3582; HUMMR; hypoxia up-regulated mitochondrial movement regulator protein; MGARP; mitochondria localized glutamic acid rich protein; mitochondria-localized glutamic acid-rich protein; OSAP; ovary-specific acidic protein; Protein MGARP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84709 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TDB4 | |
| MGARP | |
| Synthetic peptide directed towards the C-terminal of human MGARP | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction