Learn More
MGAM Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22195-1-AP-150UL
This item is not returnable.
View return policy
Description
The MGAM antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human MGAM. This antibody recognizes human, mouse antigen. The MGAM antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| MGAM | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 8972 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 447 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |