missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium, AZ10A, M11101, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Magnesium, AZ10A, M11101, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Beryllium/Iron/Nickel/Tin/Lead/Silicon/Copper/Zinc/Manganese/Aluminium |
| CAS | 7439-95-4 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Beryllium 0.00036 %/Iron 0.0034 %/Nickel 0.0041 %/Tin 0.0195 %/Lead 0.02 %/Silicon 0.062 %/Copper 0.084 %/Zinc 0.235 %/Manganese 0.377 %/Aluminium 1.083 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction