missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium, Mg/Mn, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Magnesium, Mg/Mn, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Cadmium/Iron/Lead/Tin/Nickel/Silver/Silicon/Copper/Zinc/Aluminium/Manganese/Calcium |
| CAS | 7439-95-4 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0009 %/Iron 0.0019 %/Lead 0.002 %/Tin 0.0026 %/Nickel 0.0035 %/Silver 0.0089 %/Silicon 0.019 %/Copper 0.0203 %/Zinc 0.0243 %/Aluminium 0.045 %/Manganese 1.76 %/Calcium (0.0016) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction