missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium, Mg/Al/Zn, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Magnesium, Mg/Al/Zn, Ø45x20mm
Specifications
Specifications
| Chemical Name or Material | Nickel/Beryllium/Titanium/Lead/Tin/Iron/Cadmium/Silicon/Manganese/Copper/Calcium/Zinc/Aluminium/Silver/Zirconium |
| CAS | 7439-95-4 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Nickel 0.0019 %/Beryllium 0.003 %/Titanium 0.003 %/Lead 0.004 %/Tin 0.005 %/Iron 0.007 %/Cadmium 0.011 %/Silicon 0.011 %/Manganese 0.012 %/Copper 0.021 %/Calcium 0.029 %/Zinc 0.6 %/Aluminium 3.21 %/Silver (0.002) %/Zirconium <0.001 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction