missing translation for 'onlineSavingsMsg'
Learn More

MG-132, MedChemExpress

Product Code. 16486136
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486136 100 mg 100mg
16445598 10 mg 10mg
16431767 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486136 Supplier Medchem Express Supplier No. HY13259
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MG-132 (Z-Leu-Leu-Leu-al) is a potent proteasome and calpain inhibitor with IC50s of 100 nM and 1.2 μM, respectively. MG-132 effectively blocks the proteolytic activity of the 26S proteasome complex. MG-132, a peptide aldehyde, also is an autophagy activator. MG-132 also induces apoptosis.

MG-132 (Z-Leu-Leu-Leu-al) is a potent proteasome and calpain inhibitor with IC50s of 100 nM and 1.2 μM, respectively. MG-132 effectively blocks the proteolytic activity of the 26S proteasome complex. MG-132, a peptide aldehyde, also is an autophagy activator. MG-132 also induces apoptosis.

Specifications

Chemical Name or Material MG-132
Physical Form Solid
CAS 133407-82-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C26H41N3O5
For Use With (Application) Cancer-Kinase/protease
Synonym Z-Leu-Leu-Leu-al MG132
Solubility Information DMSO : 16.67 mg/mL (35.05 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(OCC1=CC=CC=C1)N[C@H](C(N[C@@H](CC(C)C)C(N[C@H](C([H])=O)CC(C)C)=O)=O)CC(C)C
Molecular Weight (g/mol) 475.62
Formula Weight 475.62
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.